Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Bromomethyl)pyridine Hydrobromide 98.0+%, TCI America™
SDP

Catalog No. B217025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B217025G 25 g
B21705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B217025G Supplier TCI America Supplier No. B217025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4916-55-6
Molecular Formula C6H7Br2N
Molecular Weight (g/mol) 252.937
MDL Number MFCD01321314
InChI Key FNHPUOJKUXFUKN-UHFFFAOYSA-N
Synonym 3-bromomethyl pyridine hydrobromide, 3-picolyl bromide hydrobromide, 3-bromomethylpyridine hydrobromide, 3-bromomethyl-pyridine hydrobromide, pyridine, 3-bromomethyl-, hydrobromide, pubchem17664, acmc-209kf1, 3-brommethyl pyridine-hbr, 3-bromomethyl pyridine, bromide, 3-bromomethyl pyridinehydrobromide
PubChem CID 12707037
IUPAC Name 3-(bromomethyl)pyridine;hydrobromide
SMILES C1=CC(=CN=C1)CBr.Br

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 252.94
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Bromomethyl)pyridine Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.