Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophenacyl Bromide 98.0+%, TCI America™
SDP

Catalog No. B412525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B412525G 25 g
B41255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B412525G Supplier TCI America Supplier No. B412525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18523-22-3
Molecular Formula C8H6Br2O
Molecular Weight (g/mol) 277.943
MDL Number MFCD00082698
InChI Key MZBXSQBQPJWECM-UHFFFAOYSA-N
Synonym 3-bromophenacyl bromide, 2,3'-dibromoacetophenone, 2-bromo-1-3-bromophenyl ethanone, ethanone, 2-bromo-1-3-bromophenyl, 2-bromo-1-3-bromophenyl ethan-1-one, 3-bromophenacylbromide, 2-bromo-1-3-bromo-phenyl-ethanone, 2-bromo-3'-bromoacetophenone, 2-bromo-1-3-bromophenyl-1-ethanone
PubChem CID 519585
IUPAC Name 2-bromo-1-(3-bromophenyl)ethanone
SMILES C1=CC(=CC(=C1)Br)C(=O)CBr

Specifications

Melting Point 52°C
Quantity 25 g
UN Number 3261
Formula Weight 277.94
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromophenacyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.