Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophenol, 98%

Catalog No. AAA1484918
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1484918 50 g
AAA1484909 10 g
AAA1484930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1484918 Supplier Thermo Scientific Chemicals Supplier No. A1484918
Add to Cart

CAS: 591-20-8 | C6H5BrO | 173.01 g/mol

3-Bromophenol acts as an enzyme inhibitor. It is used in the preparation of 3-bromophenyl ester by reaction with benzoyl chloride in presence of triethylamine as a catalyst. It plays an important role as an intermediate in the manufacture of pharmaceuticals, agrochemicals and dyestuff and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromophenol acts as an enzyme inhibitor. It is used in the preparation of 3-bromophenyl ester by reaction with benzoyl chloride in presence of triethylamine as a catalyst. It plays an important role as an intermediate in the manufacture of pharmaceuticals, agrochemicals and dyestuff and in organic synthesis.

Solubility
Soluble in water, alkali, ethanol, ether and chloroform. Slightly soluble in carbon tetra chloride.

Notes
Light and air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-20-8
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.01
MDL Number MFCD00002253
InChI Key MNOJRWOWILAHAV-UHFFFAOYSA-N
Synonym m-bromophenol, phenol, 3-bromo, 3-bromo phenol, 3-bromo-phenol, phenol, m-bromo, unii-vmu0x6956y, meta bromophenol, meta-bromophenol, 3 bromophenol, 3-bromanylphenol
PubChem CID 11563
IUPAC Name 3-bromophenol
SMILES OC1=CC=CC(Br)=C1

Specifications

Melting Point 28°C to 32°C
Density 1.63
Boiling Point 235°C to 236°C
Flash Point >110°C (230°F)
Refractive Index 1.5972
Quantity 50 g
Beilstein 1853950
Sensitivity Light sensitive
Merck Index 14,1428
Solubility Information Soluble in water,alkali,ethanol,ether and chloroform. Slightly soluble in carbon tetra chloride.
Formula Weight 173.01
Percent Purity 98%
Chemical Name or Material 3-Bromophenol
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-706-5
RTECSNumber SJ7874900
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.