Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Bromophenyl)diphenylphosphine Oxide 98.0+%, TCI America™
SDP

Catalog No. B55301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55301G 1 g
B5530200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55301G Supplier TCI America Supplier No. B55301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10212-04-1
Molecular Formula C18H14BrOP
Molecular Weight (g/mol) 357.187
InChI Key GZZLAPUQZCXKKT-UHFFFAOYSA-N
PubChem CID 85529707
IUPAC Name 1-bromo-3-diphenylphosphorylbenzene
SMILES C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)C3=CC(=CC=C3)Br

Specifications

Melting Point 95°C
Color White
Quantity 1 g
Formula Weight 357.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (3-Bromophenyl)diphenylphosphine Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.