Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Bromophenylethynyl)trimethylsilane, 98%

Catalog No. AAH6602703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6602703 1 g
AAH6602706 5 g
AAH6602714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6602703 Supplier Thermo Scientific Chemicals Supplier No. H6602703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3989-13-7 | C11H13BrSi | 253.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3989-13-7
Molecular Formula C11H13BrSi
Molecular Weight (g/mol) 253.214
MDL Number MFCD01863664
InChI Key BXXMHQYHYOTTPM-UHFFFAOYSA-N
Synonym 3-bromophenylethynyl trimethylsilane, 2-3-bromophenyl ethynyl trimethylsilane, 3-bromophenyl ethynyl trimethylsilane, acmc-20apoa, 2-3-bromophenyl ethynyl-trimethyl-silane, 3-bromophenyltrimethylsilylacetylene, 1-bromo-3trimethylsilylethynylbenzene, 3-trimethylsilylethynyl bromobenzene, 1-bromo-3-trimethylsilylethynylbenzene, 1-trimethylsilylethynyl-3-bromobenzene
PubChem CID 3683324
IUPAC Name 2-(3-bromophenyl)ethynyl-trimethylsilane
SMILES C[Si](C)(C)C#CC1=CC(=CC=C1)Br

Specifications

Color Colorless to Yellow
Density 1.189
Boiling Point 80°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5555
Linear Formula BrC{6}H{4}C≡CSi(CH{3}){3}
Quantity 1 g
Formula Weight 253.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (3-Bromophenylethynyl)trimethylsilane
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement H315-H319-H335; Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.