Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophthalide 98.0+%, TCI America™
SDP

Catalog No. B106825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B106825G 25 g
B10685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B106825G Supplier TCI America Supplier No. B106825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6940-49-4
Molecular Formula C8H5BrO2
Molecular Weight (g/mol) 213.03
MDL Number MFCD00040520
InChI Key CLMSHAWYULIVFQ-UHFFFAOYSA-N
Synonym 3-Bromo-1(3H)-isobenzofuranone
PubChem CID 96218
IUPAC Name 3-bromo-3H-2-benzofuran-1-one
SMILES C1=CC=C2C(=C1)C(OC2=O)Br

Specifications

Melting Point 86°C
Boiling Point 138°C
Quantity 25 g
Formula Weight 213.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromophthalide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.