Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromosalicylaldehyde 98.0+%, TCI America™
SDP

Catalog No. B39221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39221G 1 g
B39225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39221G Supplier TCI America Supplier No. B39221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1829-34-1
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00016587
InChI Key STBGLXMINLWCNL-UHFFFAOYSA-N
PubChem CID 10910555
IUPAC Name 3-bromo-2-hydroxybenzaldehyde
SMILES OC1=C(Br)C=CC=C1C=O

Specifications

Melting Point 56°C
Quantity 1 g
Formula Weight 201.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromosalicylaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.