Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl cyanoacetate, 97%

Catalog No. AC148640010
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148640010 1 L Glass bottle
AC148642500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC148640010 Supplier Thermo Scientific Chemicals Supplier No. 148640010
Only null left
Add to Cart

CAS: 105-34-0 | C4H5NO2 | 99.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-34-0
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.09
MDL Number MFCD00001939
InChI Key ANGDWNBGPBMQHW-UHFFFAOYSA-N
Synonym methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
PubChem CID 7747
ChEBI CHEBI:51928
IUPAC Name methyl 2-cyanoacetate
SMILES COC(=O)CC#N

Specifications

Melting Point -13°C
Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 204°C to 207°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4170 to 1.419
Linear Formula NCCH2CO2CH3
Quantity 1 L
Beilstein 02, 584
Specific Gravity 1.12
Solubility Information Solubility in water: 54g/L in water (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 99.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl cyanoacetate, 97%
Show More Show Less
EINECSNumber 203-288-8
RTECSNumber AG4375000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.