Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromotriphenylamine 98.0+%, TCI America™
SDP

Catalog No. B40361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40361G 1 g
B40365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40361G Supplier TCI America Supplier No. B40361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78600-33-6
Molecular Formula C18H14BrN
Molecular Weight (g/mol) 324.221
MDL Number MFCD06798123
InChI Key YDXLVFKTOSKBKT-UHFFFAOYSA-N
Synonym 3-Bromo-N,N-diphenylaniline
PubChem CID 16217851
IUPAC Name 3-bromo-N,N-diphenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC(=CC=C3)Br

Specifications

Melting Point 92°C
Color White-Yellow
Boiling Point 193°C
Quantity 1 g
Formula Weight 324.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromotriphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.