Learn More
Cayman Chemical 6-chloro-MDMAhydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 90019445 MG
Encompass_Preferred
A Bcr-AblT315I inhibitor (IC50 44 nM) decreases proliferation in Ba/F3 cells expressing Bcr-AblT315I (GI50 117 nM) inhibits phosphorylation of Jak2 and Stat5 in Ba/F3 cells at 1 or 5 UM
Catalog No. 50-289-2739
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.