Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Butenylmagnesium Bromide Solution

Catalog No. 419591100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-959-1100ML 100 mL
41-959-1800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 41-959-1100ML Supplier Sigma Aldrich Supplier No. 419591100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in a key diastereoselective step in the synthesis of (-)-muricatacin fro mL-tartaric acid.

Specifications

CAS 7103-09-5
Density 0.937 g/mL at 25°C
Boiling Point 65°C
Molecular Formula C4H7BrMg
Linear Formula H2C=CHCH2CH2MgBr
MDL Number MFCD00209554
Quantity 100 mL
Molecular Weight (g/mol) 159.31 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.