Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butoxyphenol 96.0+%, TCI America™
SDP

Catalog No. B070125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B070125G 25 g
B07015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B070125G Supplier TCI America Supplier No. B070125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18979-72-1
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00058976
InChI Key VGIJZDWQVCXVNL-UHFFFAOYSA-N
Synonym Resorcinol Monobutyl Ether
PubChem CID 87876
IUPAC Name 3-butoxyphenol
SMILES CCCCOC1=CC=CC(=C1)O

Specifications

Color Yellow
Boiling Point 269°C
Quantity 25 g
Formula Weight 166.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Butoxyphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.