Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butyn-2-one, 96%

Catalog No. AC330440500
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330440500 50 g Glass Bottle
AC330440025 2.5 g Glass Bottle
AC330440100 10 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC330440500 Supplier Thermo Scientific Chemicals Supplier No. 330440500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

CAS: 1423-60-5 | C4H4O | 68.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1423-60-5
Molecular Formula C4H4O
Molecular Weight (g/mol) 68.08
MDL Number MFCD00008775
InChI Key XRGPFNGLRSIPSA-UHFFFAOYSA-N
Synonym 3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch
PubChem CID 15018
ChEBI CHEBI:48060
IUPAC Name but-3-yn-2-one
SMILES CC(=O)C#C

Specifications

Density 0.8700g/mL
Boiling Point 85.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4045 to 1.4095
Linear Formula HC≡CCOCH3
Quantity 50 g
Fieser 03,38
Specific Gravity 0.87
Formula Weight 68.06
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Butyn-2-one
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 215-834-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.