Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carbamoylpyridine-2-carboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. C1508100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1508100MG 100 mg
C15081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1508100MG Supplier TCI America Supplier No. C1508100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4733-65-7
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00661289
InChI Key CGDAWEVRGAIHJH-UHFFFAOYSA-N
Synonym 3-Carbamoylpicolinic Acid, 2-Carboxypyridine-3-carboxamide
PubChem CID 10313273
IUPAC Name 3-carbamoylpyridine-2-carboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)C(=O)N

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 166.14
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Carbamoylpyridine-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.