Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Carbobenzoxyamino)-1-propanol 98.0+%, TCI America™
SDP

Catalog No. C193225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C193225G 25 g
C19325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C193225G Supplier TCI America Supplier No. C193225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34637-22-4
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.245
MDL Number MFCD01321351
InChI Key WXQCFKYWSKKNKY-UHFFFAOYSA-N
Synonym Benzyl N-(3-Hydroxypropyl)carbamate, 3-(Cbz-amino)-1-propanol, N-(3-Hydroxypropyl)carbamic Acid Benzyl Ester
PubChem CID 562256
IUPAC Name benzyl N-(3-hydroxypropyl)carbamate
SMILES C1=CC=C(C=C1)COC(=O)NCCCO

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 25 g
Formula Weight 209.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Carbobenzoxyamino)-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.