Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carboxy-4-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH6268303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6268303 1 g
AAH62683MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6268303 Supplier Thermo Scientific Chemicals Supplier No. H6268303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882679-10-9
Molecular Formula C13H16BFO4
Molecular Weight (g/mol) 266.075
MDL Number MFCD12546519
InChI Key DXOXOBZCGBHKJS-UHFFFAOYSA-N
Synonym 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxy-4-fluorophenylboronic acid, pinacol ester, 3-carboxy-4-fluorophenylboronic acid,pinacol ester, 3-carboxy-4-fluorobenzeneboronic acid pinacol ester, 2-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, benzoic acid, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 46739598
IUPAC Name 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C(=O)O

Specifications

Quantity 1 g
Formula Weight 266.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Carboxy-4-fluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.