Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine 98.0+%, TCI America™
SDP

Catalog No. C186625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C186625G 25 g
C18665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C186625G Supplier TCI America Supplier No. C186625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32943-25-2
Molecular Formula C14H12ClN
Molecular Weight (g/mol) 229.707
MDL Number MFCD01632171
InChI Key MHUXTOYYIDFXRF-UHFFFAOYSA-N
PubChem CID 118364
IUPAC Name 2-chloro-6,11-dihydro-5H-benzo[b][1]benzazepine
SMILES C1CC2=CC=CC=C2NC3=C1C=CC(=C3)Cl

Specifications

Melting Point 87°C
Quantity 25 g
Synonym 4-Chloro-2,2′-iminodibenzyl
Formula Weight 229.71
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine
EINECSNumber (5)-1096
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.