Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2,4(3H,5H)-furandione, 97%

Catalog No. AC399650100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399650100 10 g
AC399650500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399650100 Supplier Thermo Scientific Chemicals Supplier No. 399650100
Add to Cart

CAS: 4971-55-5 | C4H3ClO3 | 134.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4971-55-5
Molecular Formula C4H3ClO3
Molecular Weight (g/mol) 134.52
MDL Number MFCD00134273
InChI Key DJEJYZZJYYNADA-UHFFFAOYNA-N
Synonym 3-chloro-2,4 3h,5h-furandione, 3-chlorofuran-2,4 3h,5h-dione, 3-chloro-furan-2,4-dione, 3-chlorotetronic acid, 2,4 3h,5h-furandione, 3-chloro, 3-chloro-3,5-dihydrofuran-2,4-dione, acmc-209khp, 3-chlorofuran-2,4-dione, 3-chloranyloxolane-2,4-dione, 3-chlorotetrahydrofuran-2,4-dione
PubChem CID 4158575
IUPAC Name 3-chlorooxolane-2,4-dione
SMILES ClC1C(=O)COC1=O

Specifications

Melting Point 208°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 10 g
Formula Weight 134.52
Percent Purity 97%
Chemical Name or Material 3-Chloro-2, 4(3H, 5H)-furandione
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.