Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2,4,5,6-tetrafluorobenzoic acid, 97%, Thermo Scientific™

Catalog No. AAL2016514
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2016514 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2016514 Supplier Thermo Scientific Chemicals Supplier No. L2016514

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5360-81-6
Molecular Formula C7HClF4O2
Quantity 25g
MDL Number MFCD03001168
Beilstein 1976066
Synonym 3-chloro-2,4,5,6-tetrafluorobenzoic acid, acmc-20an7t, 3-chloro-2,4,5,6-tetrafluoro-benzoic acid, 5-chloro-2,3,4,6-tetrafluorobenzoic acid, 3-chloranyl-2,4,5,6-tetrakis fluoranyl benzoic acid
InChI Key FYAPWZAOGRABCC-UHFFFAOYSA-N
SMILES OC(=O)C1=C(F)C(F)=C(F)C(Cl)=C1F
IUPAC Name 3-chloro-2,4,5,6-tetrafluorobenzoic acid
Molecular Weight (g/mol) 228.53
PubChem CID 2773835
Formula Weight 228.54
Percent Purity 97%
Chemical Name or Material 3-Chloro-2,4,5,6-tetrafluorobenzoic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.