Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-chloromethyl-1-propene 98.0+%, TCI America™
SDP

Catalog No. C156525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C156525G 25 g
C15655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C156525G Supplier TCI America Supplier No. C156525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1871-57-4
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 124.99
MDL Number MFCD00000952
InChI Key XJFZOSUFGSANIF-UHFFFAOYSA-N
Synonym 3-chloro-2-chloromethyl prop-1-ene, 3-chloro-2-chloromethyl-1-propene, 1,1-bis chloromethyl ethylene, methallyl dichloride, 1-propene, 3-chloro-2-chloromethyl, 2-chloromethyl-3-chloropropene, 1,3-dichloro-2-methylenepropane, 3-chloro-2-chloromethyl propene, 2-methylene-1,3-dichloropropane, 2-methylenepropane-1,3-dichloride
PubChem CID 15859
IUPAC Name 3-chloro-2-(chloromethyl)prop-1-ene
SMILES ClCC(=C)CCl

Specifications

Melting Point -14°C
Color Yellow
Boiling Point 138°C
Quantity 25 g
UN Number 2929
Formula Weight 124.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-chloromethyl-1-propene
RTECSNumber UC7400000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.