Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine, 98%, Thermo Scientific™

Catalog No. AAH6472303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6472303 1 g
AAH6472306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6472303 Supplier Thermo Scientific Chemicals Supplier No. H6472303

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72537-17-8
Molecular Formula C6H2ClF4N
Molecular Weight (g/mol) 199.53
MDL Number MFCD00191918
InChI Key GDSROTVTTLUHCO-UHFFFAOYSA-N
Synonym 3-chloro-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethylpyridine, pyridine, 3-chloro-2-fluoro-5-trifluoromethyl, acmc-20aied, librarion l939, 3-chloro-2-fluoro-5 trifluoromethyl pyridine, 3-chloro-2-fluoro-5-trifluoromethyl-pyridine
PubChem CID 1268075
IUPAC Name 3-chloro-2-fluoro-5-(trifluoromethyl)pyridine
SMILES FC1=C(Cl)C=C(C=N1)C(F)(F)F

Specifications

Melting Point 16°C to 20°C (decomposition)
Color Colorless
Density 1.524 g/mL
Boiling Point 50°C to 55°C (11 mmHg)
Flash Point 52°C (126°F)
Refractive Index 1.433
Quantity 1 g
UN Number UN1993
Formula Weight 199.53
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.