Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-fluorobenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. C211725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C211725G 25 g
C21175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C211725G Supplier TCI America Supplier No. C211725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 161957-55-7
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.555
MDL Number MFCD00042506
InChI Key FCSSYEWURMTUSM-UHFFFAOYSA-N
Synonym 2-fluoro-3-chlorobenzoic acid, 3-chloro-2-fluoro benzoic acid, 3-chloro-2-fluoro-benzoic acid, 3-chloro-2-fluorobenzoicacid, 3-chloro-2-fluorbenzoic acid, pubchem1332, acmc-209dnj, intermediates-zcf02219, benzoic acid, chlorofluoro, ksc112c3l
PubChem CID 302931
IUPAC Name 3-chloro-2-fluorobenzoic acid
SMILES C1=CC(=C(C(=C1)Cl)F)C(=O)O

Specifications

Melting Point 177°C
Quantity 25 g
Formula Weight 174.56
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.