Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. C098825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C098825G 25 g
C09885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C098825G Supplier TCI America Supplier No. C098825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4771-47-5
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
InChI Key VCHSXYHBMFKRBK-UHFFFAOYSA-N
Synonym 3-chloro-2-nitrobenzoic acid, benzoic acid, 3-chloro-2-nitro, 3-chloro-2-nitrobenzoicacid, 3-chloro-2-nitro-benzoic acid, chlornitrobenzoesaure, pubchem11043, chloro-2-nitrobenzoic acid, acmc-209ka6, 3-chloro2-nitrobenzoic acid, ksc237s1n
PubChem CID 20902
SMILES C1=CC(=C(C(=C1)Cl)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 239°C
Color White-Yellow
Quantity 25 g
Formula Weight 201.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-nitrobenzoic Acid
EINECSNumber (3)-1502
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.