Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-ethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C26641G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26641G 1 g
C2664-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26641G Supplier TCI America Supplier No. C26641G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 279261-81-3
Molecular Formula C8H10BClO3
MDL Number MFCD03701556
InChI Key QZMQZKMQXURQNC-UHFFFAOYSA-N
PubChem CID 3717658
IUPAC Name (3-chloro-4-ethoxyphenyl)boronic acid

Specifications

Melting Point 239°C
Color White-Yellow
Quantity 1 g
Synonym 3-chloro-4-ethoxyphenyl boronic acid, 3-chloro-4-ethoxybenzeneboronic acid, 3-chloro-4-ethoxypheylboronic acid, 3-chloro-4-ethoxy-phenyl boronic acid, acmc-209gzs, 3-chloro-4-ethoxyphenyl boronicacid, 3-chloranyl-4-ethoxy-phenyl boronic acid, boronic acid,b-3-chloro-4-ethoxyphenyl, 3-chloro-4-ethoxyphenylboronicacid containsvaryingamountsofanhydride
SMILES B(C1=CC(=C(C=C1)OCC)Cl)(O)O
Molecular Weight (g/mol) 200.425
Formula Weight 200.43
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-ethoxyphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.