Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-hydroxybenzoic Acid Hemihydrate 98.0+%, TCI America™
SDP

Catalog No. C1128250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1128250G 250 g
C112825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1128250G Supplier TCI America Supplier No. C1128250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3964-58-7
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD00002549
InChI Key QGNLHMKIGMZKJX-UHFFFAOYSA-N
Synonym benzoic acid, 3-chloro-4-hydroxy, 3-chloro-4-hydroxy-benzoic acid, hemihydrate, pubchem13038, acmc-209vmh, 3-chloro-4hydroxybenzoic acid, 3-chloro4-hydroxybenzoic acid, 4-hydroxy-3-chlorobenzoic acid, 3-chloro-4-hydroxybenzoic scid, 3-chloro-4-hydroxy benzoic acid
PubChem CID 19860
IUPAC Name 3-chloro-4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C(Cl)=C1

Specifications

Melting Point 171°C
Color White
Quantity 250 g
Formula Weight 172.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-hydroxybenzoic Acid Hemihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.