Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-iodoaniline 95.0+%, TCI America™
SDP

Catalog No. C216725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C216725G 25 g
C21675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C216725G Supplier TCI America Supplier No. C216725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135050-44-1
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.47
MDL Number MFCD00051845
InChI Key ONZHMGRKWJMTDE-UHFFFAOYSA-N
Synonym benzenamine, 3-chloro-4-iodo, 3-chloro-4-iodobenzenamine, 4-amino-2-chloroiodobenzene, 3-chloro-4-iodophenylamine, pubchem3812, 3-chloro4-iodoaniline, zlchem 1106, acmc-209bxp, 3-chloro-4-iodo-aniline, 3-chloro-4-iodo aniline
PubChem CID 282926
IUPAC Name 3-chloro-4-iodoaniline
SMILES NC1=CC=C(I)C(Cl)=C1

Specifications

Melting Point 70°C
Color Purple-White
Quantity 25 g
Formula Weight 253.47
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-iodoaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.