Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-methoxybenzoic acid, 97%, Thermo Scientific™

Catalog No. AC458150250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458150250 25 g
AC458150050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458150250 Supplier Thermo Scientific Chemicals Supplier No. 458150250

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37908-96-6
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.6
InChI Key IBCQUQXCTOPJOD-UHFFFAOYSA-N
Synonym 3-chloro-p-anisic acid, 3-chloro-4-methoxybenzolic acid, benzoic acid, 3-chloro-4-methoxy, pubchem19130, acmc-209iw4, ksc494s0j, 3-chloro-4-methoxybenzoicacid, rarechem al bo 2400, 3-chloro-4-anisic acid, 3-chloro-4-methoxy benzoic acid
PubChem CID 169982
IUPAC Name 3-chloro-4-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)Cl

Specifications

Melting Point 217.0°C to 219.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Formula Weight 186.6
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-methoxybenzoic acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYE
EINECSNumber 253-708-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.