Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-methoxybenzylamine hydrochloride, 97%, Thermo Scientific™

Catalog No. AAH6380606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6380606 5 g
AAH6380603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6380606 Supplier Thermo Scientific Chemicals Supplier No. H6380606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41965-95-1
Molecular Formula C8H11Cl2NO
Molecular Weight (g/mol) 208.082
MDL Number MFCD01211460
InChI Key IKWWOZCEHOYKAO-UHFFFAOYSA-N
Synonym 3-chloro-4-methoxybenzylamine hydrochloride, 3-chloro-4-methoxyphenyl methanamine hydrochloride, 3-chloro-4-methoxybenzylamine hcl, 4-aminomethyl-2-chloroanisole hcl, 4-aminomethyl-2-chloroanisole hydrochloride, 1-3-chloro-4-methoxyphenyl methanamine hydrochloride, acmc-1ambw, 3-chloro-4-methoxybenzylamine.hcl, benzenemethanamine, 3-chloro-4-methoxy-, hydrochloride 1:1, chloromethoxybenzylaminehydrochloride
PubChem CID 2764287
IUPAC Name (3-chloro-4-methoxyphenyl)methanamine;hydrochloride
SMILES COC1=C(C=C(C=C1)CN)Cl.Cl

Specifications

Melting Point 250°C to 255°C
Quantity 5 g
UN Number UN2811
Formula Weight 208.09
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Chloro-4-methoxybenzylamine hydrochloride
Hazard Category H301-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.