Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-methoxybenzylamine Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. C29035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29035G 5 g
C29031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29035G Supplier TCI America Supplier No. C29035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41965-95-1
Molecular Formula C8H11Cl2NO
Molecular Weight (g/mol) 208.082
MDL Number MFCD01211460
InChI Key IKWWOZCEHOYKAO-UHFFFAOYSA-N
Synonym 3-chloro-4-methoxybenzylamine hydrochloride, 3-chloro-4-methoxyphenyl methanamine hydrochloride, 3-chloro-4-methoxybenzylamine hcl, 4-aminomethyl-2-chloroanisole hcl, 4-aminomethyl-2-chloroanisole hydrochloride, 1-3-chloro-4-methoxyphenyl methanamine hydrochloride, acmc-1ambw, 3-chloro-4-methoxybenzylamine.hcl, benzenemethanamine, 3-chloro-4-methoxy-, hydrochloride 1:1, chloromethoxybenzylaminehydrochloride
PubChem CID 2764287
IUPAC Name (3-chloro-4-methoxyphenyl)methanamine;hydrochloride
SMILES COC1=C(C=C(C=C1)CN)Cl.Cl

Specifications

Melting Point 252°C
Color White
Quantity 5 g
Formula Weight 208.08
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-methoxybenzylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.