Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-methylbenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. C246225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C246225G 25 g
C24625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C246225G Supplier TCI America Supplier No. C246225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5162-82-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00045841
InChI Key SDKUOEOJAXGCLU-UHFFFAOYSA-N
Synonym 3-chloro-p-toluic acid, p-toluic acid, 3-chloro, benzoic acid, 3-chloro-4-methyl, 3-chloro-4-methyl-benzoic acid, 3-chloro-4-methyl benzoic acid, pubchem4569, acmc-1ayqf, 4-09-00-01743 beilstein handbook reference, ksc272s4h, 3-chloro,4-methylbenzoic acid
PubChem CID 78840
IUPAC Name 3-chloro-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)Cl

Specifications

Melting Point 202°C
Quantity 25 g
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-methylbenzoic Acid
RTECSNumber XU1650000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.