Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-methylphenyl Isocyanate 97.0+%, TCI America™
SDP

Catalog No. I056325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I056325G 25 g
I05635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I056325G Supplier TCI America Supplier No. I056325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28479-22-3
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00013858
InChI Key UKTKKMZDESVUEE-UHFFFAOYSA-N
Synonym 3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison
PubChem CID 62832
IUPAC Name 2-chloro-4-isocyanato-1-methylbenzene
SMILES CC1=CC=C(C=C1Cl)N=C=O

Specifications

Color White-Yellow
Quantity 25 g
Physical Form Crystal-Lump at 20°C
UN Number 3428
Boiling Point 124°C
Formula Weight 167.59
Percent Purity ≥97.0% (GC)
Chemical Name or Material 3-Chloro-4-methylphenyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.