Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-(trifluoromethoxy)aniline 98.0+%, TCI America™
SDP

Catalog No. C252825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C252825G 25 g
C25285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C252825G Supplier TCI America Supplier No. C252825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64628-73-5
Molecular Formula C7H5ClF3NO
Molecular Weight (g/mol) 211.568
MDL Number MFCD00190126
InChI Key ZPKUUNGPBSRPRM-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethoxy aniline, benzenamine, 3-chloro-4-trifluoromethoxy, 3-chloro-4-trifluoromethoxy phenylamine, pubchem2949, acmc-1b4hb, ksc496e4r, 3-chloro4-trifluoromethoxyaniline, 3-chloro-4-trifluoromethoxy-aniline, 3-chloro-4-trifluoromethoxy-phenylamine
PubChem CID 2773843
IUPAC Name 3-chloro-4-(trifluoromethoxy)aniline
SMILES C1=CC(=C(C=C1N)Cl)OC(F)(F)F

Specifications

Color White
Quantity 25 g
Physical Form Crystal-Lump at 20°C
Formula Weight 211.57
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 3-Chloro-4-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.