Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-5-fluoroanisole 96.0+%, TCI America™
SDP

Catalog No. C22181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22181G 1 g
C22185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22181G Supplier TCI America Supplier No. C22181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 202925-08-4
Molecular Formula C7H6ClFO
Molecular Weight (g/mol) 160.57
MDL Number MFCD00070770
InChI Key XPZBNEWAZPZUHF-UHFFFAOYSA-N
PubChem CID 2779258
IUPAC Name 1-chloro-3-fluoro-5-methoxybenzene
SMILES COC1=CC(F)=CC(Cl)=C1

Specifications

Color Yellow
Boiling Point 189°C
Quantity 1 g
Synonym 3-chloro-5-fluoroanisole, benzene,1-chloro-3-fluoro-5-methoxy, 1-chloro-3-fluoro-5-methoxy-benzene, pubchem2661, 3-chloro-5-fluoroanisol, 3-fluoro-5-methoxy-1-chlorobenzene, benzene, 1-chloro-3-fluoro-5-methoxy, 1-chloranyl-3-fluoranyl-5-methoxy-benzene
Formula Weight 160.57
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-5-fluoroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.