Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-5-trifluoromethyl-o-phenylenediamine, 97%, Thermo Scientific™

Catalog No. AAB2091003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2091003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2091003 Supplier Thermo Scientific Chemicals Supplier No. B2091003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132915-80-1
Molecular Formula C7H6ClF3N2
Molecular Weight (g/mol) 210.58
MDL Number MFCD00042198
InChI Key NVNLHLRVSHJKBN-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl benzene-1,2-diamine, 3-chloro-4,5-diaminobenzotrifluoride, 1,2-diamino-3-chloro-5-trifluoromethyl benzene, 3-chloro-5-trifluoromethyl-1,2-phenylenediamine, 3-chloro-5-trifluoromethyl-1,2-benzenediamine, timtec-bb sbb003446, buttpark 75\04-15, 1,2-diamino-3-chloro-5trifluoromethylbenzene
PubChem CID 735784
IUPAC Name 3-chloro-5-(trifluoromethyl)benzene-1,2-diamine
SMILES NC1=CC(=CC(Cl)=C1N)C(F)(F)F

Specifications

Melting Point 40°C to 43°C
Quantity 1 g
UN Number UN2811
Beilstein 4744598
Formula Weight 210.59
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-trifluoromethyl-o-phenylenediamine
Hazard Category Danger
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.