Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-5-(trifluoromethyl)pyridine 97.0+%, TCI America™
SDP

Catalog No. C29511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29511G 1 g
C29515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29511G Supplier TCI America Supplier No. C29511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85148-26-1
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD00042227
InChI Key OMRCXTBFBBWTDL-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl pyridine, 3-trifluoromethyl-5-chloropyridine, pyridine, 3-chloro-5-trifluoromethyl, pubchem3552, acmc-209zee, ksc495c1f, 3-chloro-5-trifluoromethyl-pyridine, 3-chloro-5-trifluoromethyl pyridine-carbonitrile
PubChem CID 2736698
IUPAC Name 3-chloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC(Cl)=CN=C1

Specifications

Color Yellow
Boiling Point 138°C
Quantity 1 g
UN Number 1992
Formula Weight 181.54
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.