Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-L-alanine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C16125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16125G 5 g
C16121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16125G Supplier TCI America Supplier No. C16125G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 51887-89-9
Molecular Formula C3H7Cl2NO2
Molecular Weight (g/mol) 159.994
MDL Number MFCD00070398
InChI Key IENJPSDBNBGIEL-DKWTVANSSA-N
Synonym (R)-2-Amino-3-chloropropionic Acid Hydrochloride, beta-Chloro-L-alanine Hydrochloride, H-Ala(3-Cl)-OH.HCl
PubChem CID 11957493
IUPAC Name (2R)-2-amino-3-chloropropanoic acid;hydrochloride
SMILES C(C(C(=O)O)N)Cl.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 159.99
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-L-alanine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.