Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chlorobenzal chloride, 95%

Click to view available options
Quantity:
10g
50g
15145-69-4
C7H5Cl3
195.467
MFCD00082691
ACNQJGLSENYFQJ-UHFFFAOYSA-N
3-chlorobenzal chloride,1-chloro-3-dichloromethyl benzene,alpha,alpha,3-trichlorotoluene,benzene,1-chloro-3-dichloromethyl,m-chlorbenzalchlorid,3-chlorobenzalchloride,benzene, 1-chloro-3-dichloromethyl,3-chlrobenzal chloride,acmc-20an68,1-dichloromethyl-3-chlorobenzene
84803
1-chloro-3-(dichloromethyl)benzene
C1=CC(=CC(=C1)Cl)C(Cl)Cl
15145-69-4
C7H5Cl3
195.467
MFCD00082691
ACNQJGLSENYFQJ-UHFFFAOYSA-N
3-chlorobenzal chloride,1-chloro-3-dichloromethyl benzene,alpha,alpha,3-trichlorotoluene,benzene,1-chloro-3-dichloromethyl,m-chlorbenzalchlorid,3-chlorobenzalchloride,benzene, 1-chloro-3-dichloromethyl,3-chlrobenzal chloride,acmc-20an68,1-dichloromethyl-3-chlorobenzene
84803
1-chloro-3-(dichloromethyl)benzene
C1=CC(=CC(=C1)Cl)C(Cl)Cl
Odor Pungent
Boiling Point 235°C to 237°C
Quantity 10g
Flash Point >110°C (230°F)
UN Number UN3265
Formula Weight 195.48
Percent Purity 95%
Chemical Name or Material 3-Chlorobenzal chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.