Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorobenzylamine, 97%

Catalog No. AAA1407809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1407809 10 g
AAA1407818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1407809 Supplier Thermo Scientific Chemicals Supplier No. A1407809
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4152-90-3 | C7H8ClN | 141.598 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4152-90-3
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00040752
InChI Key BJFPYGGTDAYECS-UHFFFAOYSA-N
Synonym 3-chlorobenzylamine, 3-chlorophenyl methanamine, benzenemethanamine, 3-chloro, 1-3-chlorophenyl methanamine, m-chlorobenzylamine, 3-chloro-benzylamine, m-chlorobenzyl amine, 3-chlorophenyl methylamine, chembl12957, c2a
PubChem CID 77802
IUPAC Name (3-chlorophenyl)methanamine
SMILES C1=CC(=CC(=C1)Cl)CN

Specifications

Density 1.159
Boiling Point 226°C
Flash Point 98°C (208°F)
Odor Amine-like
Refractive Index 1.559
Quantity 10 g
UN Number UN2735
Beilstein 774509
Sensitivity Air Sensitive
Formula Weight 141.6
Percent Purity ≥97%
Chemical Name or Material 3-Chlorobenzylamine
Show More Show Less
missing translation for 'healthHazard1' H314-H335
missing translation for 'healthHazard2' GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
missing translation for 'healthHazard3' P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 223-985-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.