Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorobenzylsulfonyl chloride, 97%, Thermo Scientific™

Catalog No. AC454480010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454480010 1 g
AC454480050 5 g
AC454480250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC454480010 Supplier Thermo Scientific Chemicals Supplier No. 454480010

Chemical Identifiers

CAS 24974-73-0
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.09
InChI Key LXTGNVLBPVVMSL-UHFFFAOYSA-N
PubChem CID 2757802
IUPAC Name (3-chlorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC(=C1)Cl)CS(=O)(=O)Cl

Specifications

Melting Point 71.0°C to 75.0°C
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Assay Percent Range 96% min. As diethylamine derivative (HPLC)
Synonym 3-chlorophenyl methanesulfonyl chloride, 3-chloro-phenyl-methanesulfonyl chloride, chloro 3-chlorophenyl methyl sulfone, benzenemethanesulfonyl chloride, 3-chloro, pubchem5501, acmc-20an5u, 3-chlorobenzylsulfonyl chloride, 3-chlorophenyl methanesulfonylchloride, benzenemethanesulfonylchloride,3-chloro, benzenemethanesulfonylchloride, 3-chloro
Formula Weight 225.09
Percent Purity 97%
Chemical Name or Material 3-Chlorobenzylsulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.