Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Chlorocarbonyl)-2-methylphenyl Acetate 98.0+%, TCI America™
SDP

Catalog No. C253125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C253125G 25 g
C25315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C253125G Supplier TCI America Supplier No. C253125G

Chemical Identifiers

CAS 167678-46-8
Molecular Formula C10H9ClO3
Molecular Weight (g/mol) 212.63
MDL Number MFCD02094037
InChI Key COLDUSGLGQXXEJ-UHFFFAOYSA-N
Synonym 3-acetoxy-2-methylbenzoyl chloride, 3-chlorocarbonyl-2-methylphenyl acetate, 3-acetoxy-2-methylbenzoylchloride, 3-acetyloxy-2-methyl-benzoyl chloride, 3-carbonochloridoyl-2-methylphenyl acetate, acetic acid 3-chlorocarbonyl-2-methylphenyl ester, ambc, acmc-1c4md, 3-acetoxy-o-toluoyl chloride, 3-acetoxy-2-methyl-benzoyl chloride
PubChem CID 7010345
IUPAC Name 3-(carbonochloridoyl)-2-methylphenyl acetate
SMILES CC(=O)OC1=CC=CC(C(Cl)=O)=C1C

Specifications

Melting Point 36°C
Color White-Yellow
Boiling Point 148°C
Quantity 25 g
UN Number 3261
Formula Weight 212.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-(Chlorocarbonyl)-2-methylphenyl Acetate
EINECSNumber (3)-4418
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.