Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorocoumarin 98.0+%, TCI America™
SDP

Catalog No. C22975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22975G 5 g
C22971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22975G Supplier TCI America Supplier No. C22975G

Chemical Identifiers

CAS 92-45-5
Molecular Formula C9H5ClO2
Molecular Weight (g/mol) 180.59
MDL Number MFCD00024073
InChI Key CKCOPMSBJBNBCQ-UHFFFAOYSA-N
Synonym 3-chlorocoumarin, coumarin, 3-chloro, 2h-1-benzopyran-2-one, 3-chloro, 3-chloro-2h-chromen-2-one, chlorocoumarin, 3-chloro-2h-1-benzopyran-2-one, acmc-1ciat, 2h-1-benzopyran-2-one,3-chloro
PubChem CID 96133
IUPAC Name 3-chloro-2H-chromen-2-one
SMILES ClC1=CC2=CC=CC=C2OC1=O

Specifications

Melting Point 123°C
Quantity 5 g
Formula Weight 180.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chlorocoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.