Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Chloromethyl)-3-methyloxetane 98.0+%, TCI America™
SDP

Catalog No. C122925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C122925G 25 g
C12295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C122925G Supplier TCI America Supplier No. C122925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 822-48-0
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD01709552
InChI Key MCZYEFODKAZWIH-UHFFFAOYSA-N
Synonym 2-(Chloromethyl)-2-methyl-1,3-epoxypropane
PubChem CID 79112
IUPAC Name 3-(chloromethyl)-3-methyloxetane
SMILES CC1(CCl)COC1

Specifications

Color Colorless
Boiling Point 88°C
Quantity 25 g
UN Number 1993
Formula Weight 120.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Chloromethyl)-3-methyloxetane
RTECSNumber RQ6828000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.