Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorophthalic anhydride, 95+%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
117-21-5
C8H3ClO3
182.56
MFCD00023107
UERPUZBSSSAZJE-UHFFFAOYSA-N
3-chlorophthalic anhydride,4-chloroisobenzofuran-1,3-dione,3-chlorophthalic acid anhydride,unii-vo7nxt0137,1,3-isobenzofurandione, chloro,4-chloro-1,3-dihydro-2-benzofuran-1,3-dione,4-chloranyl-2-benzofuran-1,3-dione,3clpa,pubchem19464,3-chlorophthalicanhydride
67014
4-chloro-2-benzofuran-1,3-dione
C1=CC2=C(C(=C1)Cl)C(=O)OC2=O
117-21-5
C8H3ClO3
182.56
MFCD00023107
UERPUZBSSSAZJE-UHFFFAOYSA-N
3-chlorophthalic anhydride,4-chloroisobenzofuran-1,3-dione,3-chlorophthalic acid anhydride,unii-vo7nxt0137,1,3-isobenzofurandione, chloro,4-chloro-1,3-dihydro-2-benzofuran-1,3-dione,4-chloranyl-2-benzofuran-1,3-dione,3clpa,pubchem19464,3-chlorophthalicanhydride
67014
4-chloro-2-benzofuran-1,3-dione
C1=CC2=C(C(=C1)Cl)C(=O)OC2=O
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color White or Yellow
Melting Point 123°C to 126°C
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95+%
Formula Weight 182.56
Physical Form Crystalline Powder and Chunks
Percent Purity ≥95.0% (morpholine coupling)
Chemical Name or Material 3-Chlorophthalic anhydride
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.