Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropionaldehyde Diethyl Acetal, Stabilized 95%, Thermo Scientific™

Catalog No. AC109970250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC109970250 25 g
AC109971000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109970250 Supplier Thermo Scientific Chemicals Supplier No. 109970250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35573-93-4
Molecular Formula C7H15ClO2
Molecular Weight (g/mol) 166.645
InChI Key NXHONHDWVLPPCS-UHFFFAOYSA-N
Synonym 3-chloropropionaldehyde diethyl acetal, propane, 3-chloro-1,1-diethoxy, 3-chloropropionaldehyde diethylacetal, 1-chloro-3,3-diethoxypropane, .beta.-chloropropionaldehyde diethyl acetal, 3,3-diethoxypropylchloride, 3,3-diethoxypropyl chloride, 3-ethoxy-4-oxahexyl chloride, 3-chloro-1,1-diethoxy-propane, 3-chloropropanal diethyl acetal
PubChem CID 96217
IUPAC Name 3-chloro-1,1-diethoxypropane
SMILES CCOC(CCCl)OCC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Linear Formula ClCH2CH2CH(OC2H5)2
Quantity 25 g
Beilstein 01, 632
Stabilizer 0.5% CaCO3
Formula Weight 166.65
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material 3-Chloropropionaldehyde diethyl acetal, 95%
Show More Show Less
EINECSNumber 252-623-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.