Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Chloropropyl)Triethoxysilane

Catalog No. 569615100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-961-5100ML 100 mL
56-961-51L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 56-961-5100ML Supplier Sigma Aldrich Supplier No. 569615100ML
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-Chloropropyl)Triethoxysilane

Specifications

CAS 5089-70-3
Density 1.000 g/mL at 25°C
Molecular Formula C9H21ClO3Si
Linear Formula ClCH2CH2CH2Si(OCH2CH3)3
Refractive Index n20/D 1.418
MDL Number MFCD00018985
Quantity 100 mL
Molecular Weight (g/mol) 240.8 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.