Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Chloropropyl)tris(trimethylsilyloxy)silane 96.0+%, TCI America™
SDP

Catalog No. C34371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34371G 1 g
C34375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34371G Supplier TCI America Supplier No. C34371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18077-31-1
Molecular Formula C12H33ClO3Si4
Molecular Weight (g/mol) 373.18
MDL Number MFCD00054781
InChI Key MMWCHSBIWFYBBL-UHFFFAOYSA-N
Synonym (3-Chloropropyl)tris(trimethylsiloxy)silane, 3-(3-Chloropropyl)-1,1,1,5,5,5-hexamethyl-3-[(trimethylsilyl)oxy]trisiloxane
PubChem CID 2757891
IUPAC Name 4-(3-chloropropyl)-2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptane
SMILES C[Si](C)(C)O[Si](CCCCl)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 181°C
Quantity 1 g
Formula Weight 373.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (3-Chloropropyl)tris(trimethylsilyloxy)silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.