Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropylamine hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Plastic bottle
6276-54-6
C3H8ClN·HCl
130.02
MFCD00012913
IHPRVZKJZGXTBQ-UHFFFAOYSA-N
3-chloropropylamine hydrochloride,3-chloropropan-1-amine hydrochloride,3-chloropropylamine hcl,1-amino-3-chloropropane hydrochloride,3-chloro-1-propanamine hydrochloride,1-propanamine, 3-chloro-, hydrochloride,3-chloropropylammonium chloride,acmc-1b7ws,chloropropylamine hydrochloride,ksc491e7b
11469095
3-chloropropan-1-amine;hydrochloride
C(CN)CCl.Cl
6276-54-6
C3H8ClN·HCl
130.02
MFCD00012913
IHPRVZKJZGXTBQ-UHFFFAOYSA-N
3-chloropropylamine hydrochloride,3-chloropropan-1-amine hydrochloride,3-chloropropylamine hcl,1-amino-3-chloropropane hydrochloride,3-chloro-1-propanamine hydrochloride,1-propanamine, 3-chloro-, hydrochloride,3-chloropropylammonium chloride,acmc-1b7ws,chloropropylamine hydrochloride,ksc491e7b
11469095
3-chloropropan-1-amine;hydrochloride
C(CN)CCl.Cl
Water 0.5% max.
Melting Point 148°C to 150°C
Color Beige-Yellow to White
Packaging Plastic bottle
Quantity 50g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98% min. (Ionised Chloride) (Argentometry)
Linear Formula Cl(CH2)3NH2·HCl
Beilstein 04,148
Solubility Information Solubility in water: freely soluble
Formula Weight 130.02
Physical Form Crystalline Powder or Chunks
Percent Purity 98%
Chemical Name or Material 3-Chloropropylamine hydrochloride, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-474-6

TSCA : TSCA