Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorothiophene 97.0+%, TCI America™
SDP

Catalog No. C119225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C119225G 25 g
C11925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C119225G Supplier TCI America Supplier No. C119225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17249-80-8
Molecular Formula C4H3ClS
Molecular Weight (g/mol) 118.578
MDL Number MFCD00043887
InChI Key QUBJDMPBDURTJT-UHFFFAOYSA-N
Synonym thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h
PubChem CID 87017
IUPAC Name 3-chlorothiophene
SMILES C1=CSC=C1Cl

Specifications

Color Yellow
Boiling Point 139°C
Quantity 25 g
UN Number 1993
Formula Weight 118.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.