Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyano-2-fluoropyridine 98.0+%, TCI America™
SDP

Catalog No. C253825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C253825G 25 g
C25385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C253825G Supplier TCI America Supplier No. C253825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3939-13-7
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD03095082
InChI Key USIDQCCXMGJOJM-UHFFFAOYSA-N
Synonym 3-cyano-2-fluoropyridine, 2-fluoronicotinonitrile, 2-fluoro-3-cyanopyridine, 3-pyridinecarbonitrile, 2-fluoro, 2-fluoro-nicotinonitrile, 2-fluoro-3-pyridinecarbonitrile, pubchem1108, 2-fluoronicotin-onitrile, acmc-1ckgi, 2-fluoro-3-nitrylpyridine
PubChem CID 2783254
IUPAC Name 2-fluoropyridine-3-carbonitrile
SMILES C1=CC(=C(N=C1)F)C#N

Specifications

Color Green-White
Quantity 25 g
Formula Weight 122.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Cyano-2-fluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.