Learn More
3-Cyano-2-pyridone, 98%
Description
It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 224°C to 226°C |
| Quantity | 25 g |
| UN Number | UN3439 |
| Beilstein | 4661670 |
| Solubility Information | Soluble in dimethyl sulfoxide. |
| Formula Weight | 120.11 |
| Percent Purity | 98% |
| Physical Form | Solid |
| Chemical Name or Material | 3-Cyano-2-pyridone |
Safety and Handling
| Hazard Category | H302-H318 |
| Hazard Statement | GHS H Statement H302-H312-H332-H315-H319-H335 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.